Synonym:
trans-
CAS Number 106-24-1
Linear Formula (CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
Molecular Weight 154.25
Beilstein Registry Number 1722456
EC Number 203-377-1
MDL number MFCD00002917
PubChem Substance ID 24872318
Popular Documents: FTNMR (PDF)
| Related Categories | 76/768/EEC Annex VIII, Alcohols, Aliphatic, All Alphabetically Sorted, Allergenic Compounds, |
| grade | analytical standard, for terpene analysis |
| vapor density | 5.31 (vs air) |
| assay | ≥99.0% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| refractive index |
n |
|
n | |
| bp | 229-230 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?