49508 Fluka

Sinapic acid

Ultra pure, matrix substance for MALDI-MS, ≥99.5%

DOWNLOAD MSDS (PDF)

Synonym: 3,5-Dimethoxy-4-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxy-cinnamic acid, Sinapinic acid

  • CAS Number 530-59-6

  • Empirical Formula (Hill Notation) C11H12O5

  • Molecular Weight 224.21

  •  Beilstein Registry Number 2699118

  •  EC Number 208-487-3

  •  MDL number MFCD00004401

  •  PubChem Substance ID 24872946

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Mass Spectrometry, Mass Spectrometry (MS), Matrices for MALDI Analysis,
grade   matrix substance for MALDI-MS
assay   ≥99.5% (HPLC)
  ≥99.5%
mp   ~202 °C
cation traces   Al: ≤1 mg/kg
  Ba: ≤1 mg/kg
  Ca: ≤2 mg/kg
  Cd: ≤1 mg/kg
  Co: ≤1 mg/kg
  Cr: ≤1 mg/kg
  Cu: ≤1 mg/kg
  Fe: ≤1 mg/kg
  K: ≤2 mg/kg
  Li: ≤1 mg/kg
  Mg: ≤1 mg/kg
  Mn: ≤1 mg/kg
  Na: ≤2 mg/kg
  Ni: ≤1 mg/kg
  Sr: ≤1 mg/kg
  Zn: ≤1 mg/kg
suitability   in accordance for UV test
analyte suitability   dendrimers
  fullerenes
  peptides
  proteins

Price and Availability

Customers Also Viewed

2,5-Dihydroxybenzoic acid

matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure

α-Cyano-4-hydroxycinnamic acid

suitable for MALDI-TOF MS

Sinapic acid

matrix substance for MALDI-MS, ≥99.0% (T)

Sinapic acid

≥98%, powder

<I>trans</I>-Sinapic acid

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?