Synonym: (R,R)-3,3′-Dithiobis(2-aminopropionicacid)
CAS Number 56-89-3
Linear Formula [-SCH2CH(NH2)CO2H]2
Molecular Weight 240.30
Beilstein Registry Number 1728094
E Number E921
EC Number 200-296-3
MDL number MFCD00064228
| Related Categories | A-F, All Alphabetically Sorted, Amino Acids, Analytical Standards, Analytical/Chromatography, |
| grade | certified reference material |
| TraceCERT® | |
| mp | >240 °C (dec.)(lit.) |
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34. This CRM is traceable to NIST SRM.
Certified content by quantitative NMR incl. uncertainty and expiry are given in the certificate.
Download your certificate at: http://www.sigma-aldrich.
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?