EMAIL THIS PAGE TO A FRIEND

49670 Fluka

Glutaric anhydride

technical, ≥90% (NT)

  • CAS Number 108-55-4

  • Empirical Formula (Hill Notation) C5H6O3

  • Molecular Weight 114.10

  •  Beilstein Registry Number 110051

  •  EC Number 203-593-6

  •  MDL number MFCD00006679

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Anhydride Monomers, Building Blocks, Carbonyl Compounds, Carboxylic Acid Anhydrides, Chemical Synthesis,
grade   technical
assay   ≥90% (NT)
bp   150 °C/10 mmHg(lit.)
mp   50-55 °C(lit.)
  51-55 °C

Description

Other Notes

This product has been replaced by G3806-ALDRICH | Glutaric anhydride 95%

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Search for Answers, Not Just Materials

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
MA3850000
Flash Point(F) 
343.4 °F
Flash Point(C) 
173 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

AFM imaging of immobilized fibronectin: does the surface conjugation scheme affect the protein orientation/conformation? Vallières K, Chevallier P, Sarra-Bournet C, et al. Langmuir 23(19), 9745-51, (2007)

Loading...


Catalytic enantioselective desymmetrization of meso-glutaric anhydrides using a stable Ni2-Schiff base catalyst. Gopinath P, Watanabe T, and Shibasaki M Org. Lett. 14(5), 1358-61, (2012)

Loading...


DNA microarrays with PAMAM dendritic linker systems. Benters R, Niemeyer CM, Drutschmann D, et al. Nucleic Acids Res. 30(2), E10, (2002)

Loading...


Development of an immunoassay for the beta-adrenergic agonist ractopamine. Shelver WL and Smith DJ J. Immunoassay 21(1), 1-23, (2000)

Loading...


Rhodium-catalyzed enantioselective desymmetrization of meso-3,5-dimethyl glutaric anhydride: a general strategy to syn-deoxypolypropionate synthons. Cook MJ and Rovis T J. Am. Chem. Soc. 129(30), 9302-3, (2007)

Loading...


MALDI-TOF MS analysis of a library of polymerizable cyclodextrin derivatives. Bowen RL, Farahani M, Dickens SH, et al. J. Dent. Res. 79(4), 905-11, (2000)

Loading...


Disulfide- and terminal alkyne-functionalized magnetic silica particles for enrichment of azido glycopeptides. Wang S, Xie W, Zhang X, et al. Chem. Commun. (Camb.) 48(47), 5907-9, (2012)

Loading...


The acute toxicity, primary irritancy and skin sensitizing potential of glutaric anhydride. Ballantyne B, Myers RC, and Blaszcak DL Vet. Hum. Toxicol. 34(6), 493-7, (1992)

Loading...


The disposition of serum proteins as drug-carriers in mice bearing Sarcoma 180. Tanaka T, Kaneo Y, Shiramoto S, et al. Biol. Pharm. Bull. 16(12), 1270-5, (1993)

Loading...


Preparation of budesonide-dextran conjugates using glutarate spacer as a colon-targeted drug delivery system: in vitro/in vivo evaluation in induced ulcerative colitis. Varshosaz J, Emami J, Ahmadi F, et al. J. Drug Target. 19(2), 140-53, (2011)

Loading...


A glutaric acid ester as carrier system for sustained delivery of lamuvidine (3TC) dimers. Lazrek HB, Vasseur JJ, Secrist J, et al. Nucleosides Nucleotides Nucleic Acids 26(8-9), 1103-6, (2007)

Loading...


Beil. 17,V,11,9

Aldrich MSDS 1, 991:D / Corp MSDS 1 (1), 1778:D / FT-IR 2 (1), 1218:A / FT-IR 1 (1), 720:B / FT-NMR 1 (1), 1183:A / IR-Spectra (2), 372:C / IR-Spectra (3), 420:H / NMR-Reference 2 (1), 608:C / RegBook 1 (1), 839:E / Sax 6, 1478 / Sigma FT-IR 1 (2), 591:C / Structure Index 1, 130:C:5

Related Products

replaced by

Product #

Description

Add to Cart

G3806 Glutaric anhydride, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?