Synonym: Butanedioic acid
CAS Number 110-15-6
Linear Formula HOOCCH2CH2COOH
Molecular Weight 118.09
Beilstein Registry Number 1754069
E Number E363
MDL number MFCD00002789
| Related Categories | Acid, Acids and Esters, All Alphabetically Sorted, Analytical Standards, Analytical/Chromatography, |
| grade | certified reference material |
| TraceCERT® | |
| shelf life | (limited shelf life, expiry date on the label) |
| bp | 235 °C(lit.) |
| mp | 184-186 °C(lit.) |
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34. This CRM is traceable to NIST- SRM.Certified content by quantitative NMR incl. uncertainty and expiry are given in the certificate.
Download your certificate at: http://www.sigma-aldrich.
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?