Synonym:
Luteolin 7-O-β-
CAS Number 5373-11-5
Empirical Formula (Hill Notation) C21H20O11
Molecular Weight 448.38
EC Number 226-365-8
MDL number MFCD06799436
| Related Categories | Achillea, Alphabetical, Analytical Standards, Analytical/Chromatography, Capsicum, |
| grade | analytical standard |
| assay | ≥98% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?