EMAIL THIS PAGE TO A FRIEND
51155 Fluka

Nitric acid solution

25%, for argentometric titration

Purchase

Properties

Related Categories Acid Solutions, Analytical Reagents, Analytical/Chromatography, Most common compounds for volumetric titration, Nitric acid solution,
concentration   25%
ign. residue   ≤0.0005% (as SO4)
anion traces   chloride (Cl-): ≤0.5 mg/kg
  phosphate (PO43-): ≤0.5 mg/kg
  sulfate (SO42-): ≤0.5 mg/kg
cation traces   Ag: ≤0.01 mg/kg
  Al: ≤0.5 mg/kg
  As: ≤0.01 mg/kg
  Ba: ≤0.01 mg/kg
  Be: ≤0.01 mg/kg
  Bi: ≤0.1 mg/kg
  Ca: ≤0.1 mg/kg
  Cd: ≤0.01 mg/kg
  Co: ≤0.01 mg/kg
  Cr: ≤0.02 mg/kg
  Cu: ≤0.01 mg/kg
  Fe: ≤0.2 mg/kg
  Ge: ≤0.5 mg/kg
  K: ≤0.1 mg/kg
  Li: ≤0.01 mg/kg
  Mg: ≤0.1 mg/kg
  Mn: ≤0.01 mg/kg
  Mo: ≤0.02 mg/kg
  Na: ≤0.5 mg/kg
  Ni: ≤0.02 mg/kg
  Pb: ≤0.01 mg/kg
  Sr: ≤0.01 mg/kg
  Ti: ≤0.1 mg/kg
  Tl: ≤0.5 mg/kg
  V: ≤0.01 mg/kg
  Zn: ≤0.5 mg/kg
  Zr: ≤0.1 mg/kg

Description

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Corrosive to the respiratory tract.
RIDADR 
UN 2031BFORB 8 / PGII
WGK Germany 
2
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?