51228 Fluka

Tryptic Soy Broth No. 2

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: CASO Broth No. 2, Casein Soya Broth No. 2, Soybean Casein digest Broth No. 2, TSB No. 2, Tryptone Soya Broth No. 2

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Liquid Media/Broths, Bacillus, Bacteria (General Media),
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   casein peptone, 17 g/L
  dipotassium hydrogen phosphate, 2.5 g/L
  glucose, 2.5 g/L
  sodium chloride, 5 g/L
  soya peptone, 3 g/L
final pH   7.3±0.2 (25 °C)
suitability   nonselective for Bacillus
  nonselective for Bacteria (General Media)
  nonselective for Bacteroides
  nonselective for Campylobacter
  nonselective for Candida
  nonselective for Escherichia coli and Coliforms
  nonselective for Neisseria
  nonselective for Staphylococcus
  nonselective for Streptococcus

Description

Application

Highly nutrient medium for the general purpose, formulated according to USP, FDA and Eur. Phar. regulations. The growth performance is better suited for Staphylococcus aureus strains than Fluka 22092.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

for microbiology

for microbiology, ready to use media in the bottle

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?