51523 Fluka

Heneicosane

analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 629-94-7

  • Linear Formula CH3(CH2)19CH3

  • Molecular Weight 296.57

  •  Beilstein Registry Number 1748500

  •  MDL number MFCD00009346

  •  PubChem Substance ID 24873807

  • Popular Documents:  FTIR (PDF)  

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, H,
grade   analytical standard
vapor density   10.3 (vs air)
vapor pressure   <1 mmHg ( 20 °C)
assay   ≥99.5% (GC)
shelf life   (limited shelf life, expiry date on the label)
bp   100 °C/2 mmHg(lit.)
  356.1 °C/760 mmHg
mp   39-41 °C(lit.)
  39-42 °C

Price and Availability

Customers Also Viewed

Docosane

analytical standard

Eicosane
219274Eicosane

99%

Heneicosane

98%

Tricosane

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?