CAS Number 593-49-7
Linear Formula CH3(CH2)25CH3
Molecular Weight 380.73
Beilstein Registry Number 1706242
MDL number MFCD00009862
PubChem Substance ID 24873834
Popular Documents: RAMAN FTIR (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, H, |
| grade | analytical standard |
| assay | ≥98.0% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| bp | 270 °C/15 mmHg(lit.) |
| mp | 58-60 °C(lit.) |
| 61-63 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?