51750 Fluka

Heptane

purum, ≥99.0% (GC)

DOWNLOAD MSDS (PDF)
  • CAS Number 142-82-5

  • Linear Formula CH3(CH2)5CH3

  • Molecular Weight 100.20

  •  Beilstein Registry Number 1730763

  •  EC Number 205-563-8

  •  MDL number MFCD00009544

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Heptane, Solvent by Type, Solvents More...
vapor density   3.5 (vs air)
vapor pressure   40 mmHg ( 20 °C)
  83 mmHg ( 37.7 °C)
grade   purum
assay   ≥99.0% (GC)
autoignition temp.   433 °F
expl. lim.   7 %
refractive index   n20/D 1.387(lit.)
  n20/D 1.388
bp   98 °C(lit.)
mp   −91 °C(lit.)
density   0.684 g/mL at 20 °C
  0.684 g/mL at 25 °C(lit.)

Description

Other Notes

Sales restrictions may apply

Price and Availability

Customers Also Viewed

Heptane
246654Heptane

anhydrous, 99%

Heptane
51730Heptane

analytical standard

Heptane
51745Heptane

puriss. p.a., ≥99.5% (GC)

Heptane
32287Heptane

puriss. p.a., Reag. Ph. Eur., ≥99% n-heptane basis (GC)

Heptane
H2198Heptane

ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?