Synonym: 1-Heptanesulfonic acid sodium salt monohydrate
CAS Number 207300-90-1
Linear Formula CH3(CH2)5CH2SO3Na · H2O
Molecular Weight 220.26
Beilstein Registry Number 3731762
EC Number 245-210-5
MDL number MFCD00149550
PubChem Substance ID 24874012
| Related Categories | Alkyl Sulfonates, Anionic Detergents, Building Blocks, Chemical Synthesis, Detergents, |
| description | anionic |
| grade | puriss. |
| assay | ≥99.0% (T) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?