Synonym:
trans-
CAS Number 537-98-4
Linear Formula HOC6H3(OCH3)CH=CHCO2H
Molecular Weight 194.18
Beilstein Registry Number 1570363
EC Number 208-679-7
MDL number MFCD00004400
| Related Categories | A-F, Acid, All Alphabetically Sorted, Allium, Alphabetical, |
| grade | certified reference material |
| TraceCERT® | |
| mp | 168-172 °C(lit.) |
Hydroxycinnamic acid found in plant-based foods. Antioxidant.
Certified content by quantitative NMR incl. uncertainty and expiry are given in the certificate.
Download your certificate at: http://www.sigma-aldrich.
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?