• USA Home
  • 52512 - 1,1,1,3,3,3-Hexafluoro-2-propanol

EMAIL THIS PAGE TO A FRIEND

52512 Fluka

1,1,1,3,3,3-Hexafluoro-2-propanol

puriss., ≥99.0% (GC)

Synonym: HFP, Hexafluoroisopropanol

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
grade   puriss.
assay   ≥99.0% (GC)
impurities   ≤0.1% water
refractive index   n20/D 1.275(lit.)
bp   59 °C(lit.)
mp   −4 °C(lit.)
density   1.596 g/mL at 25 °C(lit.)

Description

Other Notes

Solvent with high solubilizing potential for peptides and peptide intermediates1

This product has been replaced by 105228-ALDRICH | 1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%

Price and Availability


Search for Answers, Not Just Materials

Customers Also Viewed

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2922 6.1(8) / PGII
WGK Germany 
2
RTECS 
UB6450000

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. M. Narita et al. Bull. Chem. Soc. Jpn. 61, 281, (1988)

Thioester-isocyanides: versatile reagents for the synthesis of cycle-tail peptides. Rotstein BH, Winternheimer DJ, Yin LM, et al. Chem. Commun. (Camb.) 48(31), 3775-7, (2012)

Loading...


Hexafluoroisopropanol induces amyloid fibrils of islet amyloid polypeptide by enhancing both hydrophobic and electrostatic interactions. Yanagi K, Ashizaki M, Yagi H, et al. J. Biol. Chem. 286(27), 23959-66, (2011)

Loading...


A stepwise solvent-promoted SNi reaction of α-D-glucopyranosyl fluoride: mechanistic implications for retaining glycosyltransferases. Chan J, Tang A, and Bennet AJ J. Am. Chem. Soc. 134(2), 1212-20, (2012)

Loading...


Electrostatic force microscopy of self-assembled peptide structures. Clausen CH, Dimaki M, Panagos SP, et al. Scanning 33(4), 201-7, (2011)

Loading...


Toxic role of K+ channel oxidation in mammalian brain. Cotella D, Hernandez-Enriquez B, Wu X, et al. J. Neurosci. 32(12), 4133-44, (2012)

Loading...


Sustainable release of vancomycin, gentamicin and lidocaine from novel electrospun sandwich-structured PLGA/collagen nanofibrous membranes. Chen DW, Hsu YH, Liao JY, et al. Int. J. Pharm. 430(1-2), 335-41, (2012)

Loading...


Microfabricated electrospun collagen membranes for 3-D cancer models and drug screening applications. Hartman O Biomacromolecules 10(8), 2019-32, (2009)

Loading...


Evaluation of mobile phase composition for enhancing sensitivity of targeted quantification of oligonucleotides using ultra-high performance liquid chromatography and mass spectrometry: application to phosphorothioate deoxyribonucleic acid. Chen B and Bartlett MG J. Chromatogr. A 1288, 73-81, (2013)

Loading...


Charged diphenylalanine nanotubes and controlled hierarchical self-assembly. Wang M, Du L, Wu X, et al. ACS Nano 5(6), 4448-54, (2011)

Loading...


The effect of organic modifiers on electrospray ionization charge-state distribution and desorption efficiency for oligonucleotides. Chen B, Mason SF, and Bartlett MG J. Am. Soc. Mass Spectrom. 24(2), 257-64, (2013)

Loading...


Hydrogen-bond strengthening upon photoinduced electron transfer in ruthenium-anthraquinone dyads interacting with hexafluoroisopropanol or water. Hankache J, Hanss D, and Wenger OS J. Phys. Chem. A 116(13), 3347-58, (2012)

Loading...


Drug-eluting microfibrous patches for the local delivery of rolipram in spinal cord repair. Downing TL, Wang A, Yan ZQ, et al. J. Control. Release 161(3), 910-7, (2012)

Loading...


Role of aromatic interactions in amyloid formation by islet amyloid polypeptide. Tu LH and Raleigh DP Biochemistry 52(2), 333-42, (2013)

Loading...


Silk fibroin-based scaffolds for bone regeneration. Kuboyama N, Kiba H, Arai K, et al. J. Biomed. Mater. Res. B. Appl. Biomater. 101(2), 295-302, (2013)

Loading...


Large increase of the lifetime of a charge-separated state in a molecular triad induced by hydrogen-bonding solvent. Hankache J and Wenger OS Chemistry 18(21), 6443-7, (2012)

Loading...


Effect of synthetic aβ peptide oligomers and fluorinated solvents on Kv1.3 channel properties and membrane conductance. Lioudyno MI, Broccio M, Sokolov Y, et al. PLoS ONE 7(4), e35090, (2012)

Loading...


Preparation and properties of electrospun soy protein isolate/polyethylene oxide nanofiber membranes. Xu X, Jiang L, Zhou Z, et al. ACS Appl. Mater. Interfaces 4(8), 4331-7, (2012)

Loading...


Human dental pulp progenitor cell behavior on aqueous and hexafluoroisopropanol based silk scaffolds. Zhang W, Ahluwalia IP, Literman R, et al. J. Biomed. Mater. Res. A 97(4), 414-22, (2011)

Loading...


Two fluoroalcohols as components of basic buffers for liquid chromatography electrospray ionization mass spectrometric determination of antibiotic residues. Kipper K, Herodes K, Leito I, et al. Analyst 136(21), 4587-94, (2011)

Loading...


Fluoroalcohols as novel buffer components for basic buffer solutions for liquid chromatography electrospray ionization mass spectrometry: retention mechanisms. Kipper K, Herodes K, and Leito I J. Chromatogr. A 1218(45), 8175-80, (2011)

Loading...


Determination of sevoflurane and its metabolite hexafluoroisopropanol by direct injection of human plasma into gas chromatography-tandem mass spectrometry. Kubincová J, Szabóová A, Podolec P, et al. J. Chromatogr. A 1219, 173-6, (2012)

Loading...


Hexafluoroisopropanol induces self-assembly of β-amyloid peptides into highly ordered nanostructures. Pachahara SK, Chaudhary N, Subbalakshmi C, et al. J. Pept. Sci. 18(4), 233-41, (2012)

Loading...


Production of identical retention times and mass spectra for {delta}9-tetrahydrocannabinol and cannabidiol following derivatization with trifluoracetic anhydride with 1,1,1,3,3,3-hexafluoroisopropanol*. Andrews R and Paterson S J. Anal. Toxicol. 36(1), 61-5, (2012)

Loading...


Infrared spectra, vibrational assignment, and ab initio calculations for N-bromo-hexafluoro-2-propanimine. Panikar SS, Guirgis GA, Sheehan TG, et al. Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 90, 118-24, (2012)

Loading...


Prevention of amyloid-β fibril formation using antibodies against the C-terminal region of amyloid-β1-40 and amyloid-β1-42. Montañés M, Casabona D, Sarasa L, et al. J. Alzheimers Dis. 34(1), 133-7, (2013)

Loading...


Bacterial amyloids. Zhou Y, Blanco LP, Smith DR, et al. Methods Mol. Biol. 849, 303-20, (2012)

Loading...


Application of photochemical cross-linking to the study of oligomerization of amyloidogenic proteins. Lopes DH, Sinha S, Rosensweig C, et al. Methods Mol. Biol. 849, 11-21, (2012)

Loading...


Aldrich MSDS 1, 1023:C / Corp MSDS 1 (1), 1847:C / FT-IR 1 (1), 179:A / FT-IR 2 (1), 259:A / FT-NMR 1 (1), 279:C / IR-Spectra (2), 102:D / IR-Spectra (3), 110:D / NMR-Reference 2 (1), 167:B / RegBook 1 (1), 181:D / Sax 6, 1513 / Sigma FT-IR 1 (1), 1341:C / Structure Index 1, 24:C:2 / Vapor Phase 3, 259:D

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?