52650 Fluka

Hexamethylene diisocyanate

purum, ≥98.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: 1,6-Diisocyanatohexane

  • CAS Number 822-06-0

  • Linear Formula OCN(CH2)6NCO

  • Molecular Weight 168.19

  •  Beilstein Registry Number 956709

  •  EC Number 212-485-8

  •  MDL number MFCD00002047

  •  PubChem Substance ID 24874558

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanate Monomers, Isocyanates, Materials Science,
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.453
bp   82-85 °C/0.1 mmHg
density   1.047 g/mL at 20 °C(lit.)

Price and Availability

Customers Also Viewed

Hexamethylene diisocyanate

puriss., ≥99.0% (GC)

Poly(hexamethylene diisocyanate)

viscosity 1,300-2,200 cP (25 °C)(lit.)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?