Synonym:
2-
CAS Number 51-45-6
Empirical Formula (Hill Notation) C5H9N3
Molecular Weight 111.15
Beilstein Registry Number 2012
EC Number 200-100-6
MDL number MFCD00005210
PubChem Substance ID 24877925
Popular Documents: RAMAN FTIR (PDF)
| Related Categories | Bioactive Small Molecules, Building Blocks, C3 to C8, Cell Biology, Chemical Synthesis, |
| assay | ≥97.0% (NT) |
| bp | 167 °C/0.8 mmHg(lit.) |
| mp | 83-84 °C(lit.) |
| ~84 °C | |
| storage temp. | −20°C |
| Gene Information |
human ... CA1(759), CA2(760), HRH1(3269), HRH2(3274), HRH3(11255), HRH4(59340) rat ... Hrh1(24448), Hrh3(85268) |
Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilizes Ca2+; H2 activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?