53302 Fluka

Histamine dihydrochloride

Ph Eur

DOWNLOAD MSDS (PDF)

Synonym: 2-(4-Imidazolyl)ethylamine dihydrochloride, Histamini dihydrochloridum

  • CAS Number 56-92-8

  • Empirical Formula (Hill Notation) C5H9N3 · 2HCl

  • Molecular Weight 184.07

  •  Beilstein Registry Number 3624116

  •  EC Number 200-298-4

  •  MDL number MFCD00012703

  •  PubChem Substance ID 24877934

  • Popular Documents:  FTIR (PDF)  

Properties

Related Categories Amino Acids, Building Blocks, C3 to C8, Chemical Synthesis, Core Bioreagents,
grade   Ph Eur
pharmacopeia   testing & handling conforms to Pharmacopeia
mp   249-252 °C(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Histamine

≥97.0%

Histamine

base, ≥97.0% (NT)

Histamine dihydrochloride

≥99% (TLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?