EMAIL THIS PAGE TO A FRIEND

53960 Fluka

Hydroquinone

≥99.0% (HPLC)

Synonym: 1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ

Purchase

Properties

vapor density   3.81 (vs air)
vapor pressure   1 mmHg ( 132 °C)
assay   ≥99.0% (HPLC)
autoignition temp.   930 °F
bp   285 °C(lit.)
mp   171-173 °C
  172-175 °C(lit.)
solubility   H2O: soluble50 mg/mL, clear

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Hydroquinone

meets USP testing specifications

Hydroquinone

certified reference material, TraceCERT®

Hydroquinone

United States Pharmacopeia (USP) Reference Standard

Hydroquinone

ReagentPlus®, ≥99.5%

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36/37/39-61
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
MX3500000
Flash Point(F) 
329 °F
Flash Point(C) 
165 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Chemical genetics reveals a complex functional ground state of neural stem cells Diamandis, P., et. al. Nat. Chem. Biol. 3(5), 268-273, (2007)

Loading...


Cheminformatics analysis of assertions mined from literature that describe drug-induced liver injury in different species. Denis Fourches et al Chem. Res. Toxicol. 23, 171-83, (2010)

Loading...


Alteration of the oxygen-dependent reactivity of de novo Due Ferri proteins. Reig AJ, Pires MM, Snyder RA, et al. Nature Chemistry 4(11), 900-6, (2012)

Loading...


Genetic mapping of targets mediating differential chemical phenotypes in Plasmodium falciparum. Jing Yuan et al Nat. Chem. Biol. 5, 765-71, (2009)

Loading...


Differential 12C-/13C-isotope dansylation labeling and fast liquid chromatography/mass spectrometry for absolute and relative quantification of the metabolome. Guo K and Li L Anal. Chem. 81(10), 3919-32, (2009)

Loading...


The safety of hydroquinone. Nordlund JJ, Grimes PE, and Ortonne JP J. Eur. Acad. Dermatol. Venereol. 20(7), 781-7, (2006)

Loading...


Hydroquinone regulates hemeoxygenase-1 expression via modulation of Src kinase activity through thiolation of cysteine residues. Byeon SE, Yu T, Yang Y, et al. Free Radic. Biol. Med. 57, 105-18, (2013)

Loading...


In vitro effects of aldehydes present in tobacco smoke on gene expression in human lung alveolar epithelial cells. Cheah NP, Pennings JL, Vermeulen JP, et al. Toxicol. In Vitro 27(3), 1072-81, (2013)

Loading...


Role of proton-coupled electron transfer in the redox interconversion between benzoquinone and hydroquinone. Song N, Gagliardi CJ, Binstead RA, et al. J. Am. Chem. Soc. 134(45), 18538-41, (2012)

Loading...


The toxicology of hydroquinone--relevance to occupational and environmental exposure. DeCaprio AP Crit. Rev. Toxicol. 29(3), 283-330, (1999)

Loading...


QSAR study on permeability of hydrophobic compounds with artificial membranes. Masaaki Fujikawa et al Bioorg. Med. Chem. 15, 3756-67, (2007)

Loading...


Air to lung partition coefficients for volatile organic compounds and blood to lung partition coefficients for volatile organic compounds and drugs. Michael H Abraham et al Eur. J. Med. Chem. 43, 478-85, (2008)

Loading...


Glassy carbon electrodes modified with gold nanoparticles for the simultaneous determination of three food antioxidants. Lin X, Ni Y, and Kokot S Anal. Chim. Acta 765, 54-62, (2013)

Loading...


Hydroquinone, a benzene metabolite, and leukemia: a case report and review of the literature. Regev L, Wu M, Zlotolow R, et al. Toxicol. Ind. Health 28(1), 64-73, (2012)

Loading...


Identification of a potent and noncytotoxic inhibitor of melanin production. Soonho Hwang et al Bioorg. Med. Chem. 18, 5602-9, (2010)

Loading...


A split-face evaluation of a novel pigment-lightening agent compared with no treatment and hydroquinone. Draelos ZD J. Am. Acad. Dermatol. 72(1), 105-7, (2015)

Loading...


Evaluation of in vivo mutagenicity of hydroquinone in Muta™ mice. Matsumoto M, Masumori S, Hirata-Koizumi M, et al. Mutat. Res. Genet. Toxicol. Environ. Mutagen. 775-776, 94-8, (2014)

Loading...


Synthesis of gold nanorods and their functionalization with bovine serum albumin for optical hyperthermia. Zhang L, Xia K, Bai YY, et al. J. Biomed. Nanotechnol. 10(8), 1440-9, (2014)

Loading...


Topical treatment of hyperpigmentation disorders. Rendon M and Horwitz S Ann. Dermatol. Venereol. 139 Suppl 4, S153-8, (2012)

Loading...


Efficacy of hydroquinone-free skin-lightening cream for photoaging. Dreher F, Draelos ZD, Gold MH, et al. J. Cosmet. Dermatol. 12(1), 12-7, (2013)

Loading...


4-n-butylresorcinol, a highly effective tyrosinase inhibitor for the topical treatment of hyperpigmentation. Kolbe L, Mann T, Gerwat W, et al. J. Eur. Acad. Dermatol. Venereol. 27 Suppl 1, 19-23, (2013)

Loading...


DeoxyArbutin and its derivatives inhibit tyrosinase activity and melanin synthesis without inducing reactive oxygen species or apoptosis. Chawla S, Kvalnes K, deLong MA, et al. Journal. of. Drugs in. Dermatology. 11(10), e28-34, (2012)

Loading...


In vitro inhibition of ?-carbonic anhydrase isozymes by some phenolic compounds. Sevim Beyza Oztürk Sarikaya et al Bioorg. Med. Chem. Lett. 21, 4259-62, (2011)

Loading...


Hydroquinone and its analogues in dermatology - a risk-benefit viewpoint. O'Donoghue JL J. Cosmet. Dermatol. 5(3), 196-203, (2006)

Loading...


A practical comparison of hydroquinone-containing products for the treatment of melasma. Abramovits W, Barzin S, and Arrazola P Skinmed 4(6), 371-6, (2005)

Loading...


[Protein changes in human embryonic lung fibroblasts after hydroquinone stimulation using proteomic technique]. Li X, Zhuang Z, Liu J, et al. Wei Sheng Yan Jiu 33(6), 654-7, (2004)

Loading...


[Toxicological aspects and health risks associated with hydroquinone in skin bleaching formula]. Kooyers TJ and Westerhof W Ned. Tijdschr. Geneeskd. 148(16), 768-71, (2004)

Loading...


The use of hydroquinone with facial laser resurfacing. Goldman MP J. Cutan. Laser Ther. 2(2), 73-7, (2000)

Loading...


Hypothesis: phenol and hydroquinone derived mainly from diet and gastrointestinal flora activity are causal factors in leukemia. McDonald TA, Holland NT, Skibola C, et al. Leukemia 15(1), 10-20, (2001)

Loading...


Hydroquinone. IARC Monogr. Eval. Carcinog. Risks Hum. 71 Pt 2, 691-719, (1999)

Loading...


Caspase-dependent and -independent mechanisms in apoptosis induced by hydroquinone and catechol metabolites of remoxipride in HL-60 cells. Inayat-Hussain SH, McGuinness SM, Johansson R, et al. Chem. Biol. Interact. 128(1), 51-63, (2000)

Loading...


Hydroquinone: an evaluation of the human risks from its carcinogenic and mutagenic properties. McGregor D Crit. Rev. Toxicol. 37(10), 887-914, (2007)

Loading...


Effect of the benzene metabolite, hydroquinone, on interleukin-1 secretion by human monocytes in vitro. Carbonnelle P, Lison D, Leroy JY, et al. Toxicol. Appl. Pharmacol. 132(2), 220-6, (1995)

Loading...


The safety of hydroquinone: a dermatologist's response to the 2006 Federal Register. Levitt J J. Am. Acad. Dermatol. 57(5), 854-72, (2007)

Loading...


Hydroquinone increases 5-hydroxymethylcytosine formation through ten eleven translocation 1 (TET1) 5-methylcytosine dioxygenase. Coulter JB, O'Driscoll CM, and Bressler JP J. Biol. Chem. 288(40), 28792-800, (2013)

Loading...


Photoelectrochemical sensing for hydroquinone based on porphyrin-functionalized Au nanoparticles on graphene. Hu Y, Xue Z, He H, et al. Biosens. Bioelectron. 47, 45-9, (2013)

Loading...


p38 MAPK/PP2Acα/TTP pathway on the connection of TNF-α and caspases activation on hydroquinone-induced apoptosis. Liu WH, Chou WM, and Chang LS Carcinogenesis 34(4), 818-27, (2013)

Loading...


Carbonic anhydrase inhibitors: inhibition of mammalian isoforms I-XIV with a series of substituted phenols including paracetamol and salicylic acid. Alessio Innocenti et al Bioorg. Med. Chem. 16, 7424-8, (2008)

Loading...


Hydroquinone modified chitosan/carbon film electrode for the selective detection of ascorbic acid. Jirimali HD, Nagarale RK, Saravanakumar D, et al. Carbohydr. Polym. 92(1), 641-4, (2013)

Loading...


Hydroquinone therapy for post-inflammatory hyperpigmentation secondary to acne: not just prescribable by dermatologists. Chandra M, Levitt J, and Pensabene CA Acta Derm. Venereol. 92(3), 232-5, (2012)

Loading...


S-glutathionyl-(chloro)hydroquinone reductases: a new class of glutathione transferases functioning as oxidoreductases. Belchik SM and Xun L Drug Metab. Rev. 43(2), 307-16, (2011)

Loading...


Cellular apoptosis and cytotoxicity of phenolic compounds: a quantitative structure-activity relationship study. Cynthia D Selassie et al J. Med. Chem. 48, 7234-42, (2005)

Loading...


Evaluation of anti-pigmentary effect of synthetic sulfonylamino chalcone. Woo Duck Seo et al Eur. J. Med. Chem. 45, 2010-7, (2010)

Loading...


Ultrathin CdSe nanosheets: synthesis and application in simultaneous determination of catechol and hydroquinone. Cao X, Cai X, Feng Q, et al. Anal. Chim. Acta 752, 101-5, (2012)

Loading...


Time-resolved photoelectron imaging of excited state relaxation dynamics in phenol, catechol, resorcinol, and hydroquinone. Livingstone RA, Thompson JO, Iljina M, et al. J. Chem. Phys. 137(18), 184304, (2012)

Loading...


Tyrosinase-inhibitory constituents from the twigs of Cinnamomum cassia. Tran Minh Ngoc et al J. Nat. Prod. 72, 1205-8, (2009)

Loading...


Label-free electrochemical immunosensor for prostate-specific antigen based on silver hybridized mesoporous silica nanoparticles. Wang H, Zhang Y, Yu H, et al. Anal. Biochem. 434(1), 123-7, (2013)

Loading...


Carbonic anhydrase inhibitors: Inhibition of the new membrane-associated isoform XV with phenols. Alessio Innocenti et al Bioorg. Med. Chem. Lett. 18, 3593-6, (2008)

Loading...


Skin lightening preparations and the hydroquinone controversy. Draelos ZD Dermatol. Ther. 20(5), 308-13, (2007)

Loading...


Hydroxylation of phenol to hydroquinone catalyzed by a human myeloperoxidase-superoxide complex: possible implications in benzene-induced myelotoxicity. Subrahmanyam VV, Kolachana P, and Smith MT Free Radic. Res. Commun. 15(5), 285-96, (1991)

Loading...


Pyronane monoterpenoids from the fruit of Gardenia jasminoides. Quan Cheng Chen et al J. Nat. Prod. 71, 995-9, (2008)

Loading...


Clinical efficacy and safety of a multimodality skin brightener composition compared with 4% hydroquinone. Makino ET, Mehta RC, Garruto J, et al. Journal. of. Drugs in. Dermatology. 12(3), s21-6, (2013)

Loading...


Evaluation of a hydroquinone-free skin brightening product using in vitro inhibition of melanogenesis and clinical reduction of ultraviolet-induced hyperpigmentation. Makino ET, Mehta RC, Banga A, et al. Journal. of. Drugs in. Dermatology. 12(3), s16-20, (2013)

Loading...


Analogues of N-hydroxy-N'-phenylthiourea and N-hydroxy-N'-phenylurea as inhibitors of tyrosinase and melanin formation. Marc Criton et al Bioorg. Med. Chem. Lett. 18, 3607-10, (2008)

Loading...


Convenient QSAR model for predicting the complexation of structurally diverse compounds with beta-cyclodextrins. Alfonso Pérez-Garrido et al Bioorg. Med. Chem. 17, 896-904, (2009)

Loading...


Comparison of the repair of DNA damage induced by the benzene metabolites hydroquinone and p-benzoquinone: a role for hydroquinone in benzene genotoxicity. Gaskell M, McLuckie KI, and Farmer PB Carcinogenesis 26(3), 673-80, (2005)

Loading...


Clinical efficacy and safety of a multimodality skin brightener composition compared with 4% hydroquinone. Makino ET, Herndon JH, Sigler ML, et al. Journal. of. Drugs in. Dermatology. 11(12), 1478-82, (2012)

Loading...


Effects of 3-(4'-geranyloxy-3'-methoxyphenyl)-2-trans propenoic acid and its ester derivatives on biofilm formation by two oral pathogens, Porphyromonas gingivalis and Streptococcus mutans. Charles Bodet et al Eur. J. Med. Chem. 43, 1612-20, (2008)

Loading...


Melanin synthesis inhibitors from Lespedeza cyrtobotrya. Maya Mori-Hongo et al J. Nat. Prod. 72, 63-71, (2009)

Loading...


Degradation of paracetamol by Pseudomonas aeruginosa strain HJ1012. Hu J, Zhang LL, Chen JM, et al. J. Environ. Sci. Health. A. Tox. Hazard. Subst. Environ. Eng. 48(7), 791-9, (2013)

Loading...


Synthesis and anti-inflammatory activity of 3-(4'-geranyloxy-3'-methoxyphenyl)-2-trans propenoic acid and its ester derivatives. Francesco Epifano et al Bioorg. Med. Chem. Lett. 17, 5709-14, (2007)

Loading...


Cytotoxicity of dental compounds towards human oral squamous cell carcinoma and normal oral cells. Koh T, Machino M, Murakami Y, et al. In Vivo 27(1), 85-95, (2013)

Loading...


Syntheses of hydroxy substituted 2-phenyl-naphthalenes as inhibitors of tyrosinase. Suhee Song et al Bioorg. Med. Chem. Lett. 17, 461-4, (2007)

Loading...


Melanin synthesis inhibitors from Lespedeza floribunda. Maya Mori-Hongo et al J. Nat. Prod. 72, 194-203, (2009)

Loading...


Electrochemical microdevice for on-site determination of rice freshness. Koyachi E, Kojima K, Qiu X, et al. Biosens. Bioelectron. 42, 640-5, (2013)

Loading...


Hydroquinone for skin lightening: safety profile, duration of use and when should we stop? Tse TW J. Dermatolog. Treat. 21(5), 272-5, (2010)

Loading...


Hydroquinone-induced exogenous ochronosis in Chinese--two case reports and a review. Tan SK, Sim CS, and Goh CL Int. J. Dermatol. 47(6), 639-40, (2008)

Loading...


Discovery of novel SERCA inhibitors by virtual screening of a large compound library. Christopher Elam et al Eur. J. Med. Chem. 46, 1512-23, (2011)

Loading...


Reactivity and biological activity of the marine sesquiterpene hydroquinone avarol and related compounds from sponges of the order Dictyoceratida. Sladić D and Gasić MJ Molecules 11(1), 1-33, (2006)

Loading...


A newly synthesized, potent tyrosinase inhibitor: 5-(6-hydroxy-2-naphthyl)-1,2,3-benzenetriol. Jehun Choi et al Bioorg. Med. Chem. Lett. 20, 4882-4, (2010)

Loading...


Studies on depigmenting activities of dihydroxyl benzamide derivatives containing adamantane moiety. Ho Sik Rho et al Bioorg. Med. Chem. Lett. 19, 1532-3, (2009)

Loading...


Hydroquinone-induced miR-122 down-regulation elicits ADAM17 up-regulation, leading to increased soluble TNF-α production in human leukemia cells with expressed Bcr/Abl. Chen YJ and Chang LS Biochem. Pharmacol. 86(5), 620-31, (2013)

Loading...


The effects of Sophora angustifolia and other natural plant extracts on melanogenesis and melanin transfer in human skin cells. Singh SK, Baker R, Wibawa JI, et al. Exp. Dermatol. 22(1), 67-9, (2013)

Loading...


Changes in DNA methylation of erythroid-specific genes in K562 cells exposed to phenol and hydroquinone. Li Y, Wu XR, Li XF, et al. Toxicology 312, 108-14, (2013)

Loading...


An ortho-carbonyl substituted hydroquinone derivative is an anticancer agent that acts by inhibiting mitochondrial bioenergetics and by inducing G₂/M-phase arrest in mammary adenocarcinoma TA3. Urra FA, Martínez-Cifuentes M, Pavani M, et al. Toxicol. Appl. Pharmacol. 267(3), 218-27, (2013)

Loading...


The vitamin K oxidoreductase is a multimer that efficiently reduces vitamin K epoxide to hydroquinone to allow vitamin K-dependent protein carboxylation. Rishavy MA, Hallgren KW, Wilson LA, et al. J. Biol. Chem. 288(44), 31556-66, (2013)

Loading...


[Topical treatment of hyperpigmentation disorders]. Rendon M and Horwitz S Ann. Dermatol. Venereol. 139 Suppl 3, S102-7, (2012)

Loading...


Hydroquinone concentrations in skin lightening creams. Boyle J and Kennedy CT Br. J. Dermatol. 114(4), 501-4, (1986)

Loading...


Final amended safety assessment of hydroquinone as used in cosmetics. Andersen FA, Bergfeld WF, Belsito DV, et al. Int. J. Toxicol. 29(6 Suppl), 274S-87, (2010)

Loading...


Percutaneous absorption of hydroquinone in humans: effect of 1-dodecylazacycloheptan-2-one (azone) and the 2-ethylhexyl ester of 4-(dimethylamino)benzoic acid (Escalol 507). Bucks DA, McMaster JR, Guy RH, et al. J. Toxicol. Environ. Health 24(3), 279-89, (1988)

Loading...


The percutaneous absorption of hydroquinone (HQ) through rat and human skin in vitro. Barber ED, Hill T, and Schum DB Toxicol. Lett. 80(1-3), 167-72, (1995)

Loading...


Resveratrol inhibited hydroquinone-induced cytotoxicity in mouse primary hepatocytes. Wang DH, Ootsuki Y, Fujita H, et al. Int. J. Environ. Res. Public Health 9(9), 3354-64, (2012)

Loading...


Laccase immobilized onto poly(GMA-MAA) microspheres for p-benzenediol removal from wastewater. Li C, Lou Y, Wan Y, et al. Water Sci. Technol. 67(10), 2287-93, (2013)

Loading...


Successful treatment of hydroquinone-resistant melasma using topical methimazole. Malek J, Chedraoui A, Nikolic D, et al. Dermatol. Ther. 26(1), 69-72, (2013)

Loading...


Acne and skin bleaching in Lomé, Togo. Kombaté K, Mouhari-Toure A, Saka B, et al. Int. J. Dermatol. 51 Suppl 1, 27-9, 30-2, (2012)

Loading...


Improving a self-curing dental resin by eliminating oxygen, hydroquinone and water from its curing process. Keh ES, Hayakawa I, Takahashi H, et al. Dent. Mater. J. 21(4), 373-82, (2002)

Loading...


Comparative study of hydroquinone-free and hydroquinone-based hyperpigmentation regimens in treating facial hyperpigmentation and photoaging. Fabi SG and Goldman MP Journal. of. Drugs in. Dermatology. 12(3), S32-7, (2013)

Loading...


Comparison of a skin-lightening cream targeting melanogenesis on multiple levels to triple combination cream for melasma. Monheit GD and Dreher F Journal. of. Drugs in. Dermatology. 12(3), 270-4, (2013)

Loading...


Tolerance and efficacy of a product containing ellagic and salicylic acids in reducing hyperpigmentation and dark spots in comparison with 4% hydroquinone. Dahl A, Yatskayer M, Raab S, et al. Journal. of. Drugs in. Dermatology. 12(1), 52-8, (2013)

Loading...


Treatment of melasma and the use of intense pulsed light: a review. Zaleski L, Fabi S, and Goldman MP Journal. of. Drugs in. Dermatology. 11(11), 1316-20, (2012)

Loading...


Induced differentiation of HL-60 promyelocytic leukemia cells to monocyte/macrophages is inhibited by hydroquinone, a hematotoxic metabolite of benzene. Oliveira NL and Kalf GF Blood 79(3), 627-33, (1992)

Loading...


Risk assessment of free hydroquinone derived from Arctostaphylos Uva-ursi folium herbal preparations. de Arriba SG, Naser B, and Nolte KU Int. J. Toxicol. 32(6), 442-53, (2013)

Loading...


Treating epidermal melasma with a 4% hydroquinone skin care system plus tretinoin cream 0.025%. Grimes P and Watson J Cutis. 91(1), 47-54, (2013)

Loading...


Hydroquinone: environmental pollution, toxicity, and microbial answers. Enguita FJ and Leitão AL Biomed Res. Int. 2013, 542168, (2013)

Loading...


Application of physiologically-based toxicokinetic modelling in oral-to-dermal extrapolation of threshold doses of cosmetic ingredients. Gajewska M, Worth A, Urani C, et al. Toxicol. Lett. 227(3), 189-202, (2014)

Loading...


α-Tocopheryl succinate pre-treatment attenuates quinone toxicity in prostate cancer PC3 cells. Bellezza I, Grottelli S, Gatticchi L, et al. Gene 539(1), 1-7, (2014)

Loading...


Nonextractable residue formation of sulfonamide antimicrobials: new insights from soil incubation experiments. Gulkowska A, Thalmann B, Hollender J, et al. Chemosphere 107, 366-72, (2014)

Loading...


Combination of fractional erbium-glass laser and topical therapy in melasma resistant to triple-combination cream. Tourlaki A, Galimberti MG, Pellacani G, et al. J. Dermatolog. Treat. 25(3), 218-22, (2014)

Loading...


Merck 14,4808

Beil. 6,IV,5712

Fieser 5,341

Aldrich MSDS 1, 1050:A / Corp MSDS 1 (1), 1900:B / FT-IR 2 (2), 1892:A / FT-IR 1 (1), 1107:D / FT-NMR 1 (2), 304:C / IR-Spectra (2), 594:F / IR-Spectra (3), 666:C / NMR-Reference 2 (1), 902:B / RegBook 1 (1), 1301:A / Sax 6, 1553 / Sigma FT-IR 1 (2), 618:B / Structure Index 1, 198:B:6 / Vapor Phase 3, 1041:B

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?