EMAIL THIS PAGE TO A FRIEND

54194 Fluka

2-Adamantanol

purum, ≥98.0% (GC)

Synonym: 2-Hydroxyadamantane

  • CAS Number 700-57-2

  • Empirical Formula (Hill Notation) C10H16O

  • Molecular Weight 152.23

  •  Beilstein Registry Number 2498536

  •  EC Number 211-846-7

  •  MDL number MFCD00074744

  •  PubChem Substance ID 24878569

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

grade   purum
assay   ≥98.0% (GC)
mp   258-262 °C(lit.)
  258-262 °C

Price and Availability

Customers Also Viewed

1-Adamantanol

ReagentPlus®, 99%

2-Adamantanol

97%

Adamantane

≥99%

Safety & Documentation

Safety Information

WGK Germany 
3

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

FT-IR 1 (1), 170:D / FT-IR 2 (1), 240:A / FT-NMR 1 (1), 255:C / IR-Spectra (2), 97:B / IR-Spectra (3), 104:G / NMR-Reference 2 (1), 159:D / RegBook 1 (1), 169:F / Structure Index 1, 22:C:1 / Vapor Phase 3, 247:C

Related Products

replaced by

Product #

Description

Add to Cart

153826 2-Adamantanol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?