Synonym: Dodecylthiol functionalized gold nanoparticles
EC Number 203-625-9
MDL number MFCD09953506
| Related Categories | 79: Au, Biomedical Materials, Gold Nanoparticles, Materials Science, Nanomaterials, |
| description | nanoparticles |
| assay | 0.6-0.9% solid material basis |
| form | dispersion |
| concentration | 0.01% HAuCl4 in toluene |
| particle size | 3-6 nm |
weak absorption peak at 520 nm
Useful for the self-assembly of gold monolayers for nanoelectronics.1
Air-stable
Surface stabilized with tetraoctylammonium bromide
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?