MDL number MFCD00001484
PubChem Substance ID 24879219
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, M, MEA - MES, |
| grade | NMR reference standard |
| concentration | 30 wt. % in chloroform-d (99.8 atom % D) |
| NMR tube O.D. × L | 5 mm × 8 in. |
APT and DEPT demonstration
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?