55154 Fluka

Octadecyl 7-hydroxycoumarin-3-carboxylate

for fluorescence, ≥98.0% (TLC)

DOWNLOAD MSDS (PDF)

Synonym: 7-Hydroxycoumarin-3-carboxylic acid octadecyl ester, Umbelliferone-3-carboxylic acid octadecyl ester

  • Empirical Formula (Hill Notation) C28H42O5

  • Molecular Weight 458.63

  •  MDL number MFCD00037567

  •  PubChem Substance ID 24879224

Properties

Related Categories Alphabetical, Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, Detection,
grade   for fluorescence
assay   ≥98.0% (TLC)
mp   114-116 °C(lit.)
solubility   DMF: soluble
  DMSO: soluble
  alcohols: soluble
fluorescence   λex 390 nm; λem 445 nm in 0.1 M Tris pH 9.0
storage temp.   2-8°C

Price and Availability

Customers Also Viewed

7-<WBR>Hydroxycoumarin-<WBR>3-<WBR>carboxylic acid

suitable for fluorescence, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?