55876 Fluka

Adonitol Disks

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories Discs and Strips, Identification Tests & Reagents, Microbiology More...
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
storage temp.   2-8°C

Description

Application

Used to differentiate bacteria on the basis of carbohydrate fermentation abilities

Price and Availability

Customers Also Viewed

Adonitol

≥99%

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?