Synonym:
3-
CAS Number 874-24-8
Empirical Formula (Hill Notation) C6H5NO3
Molecular Weight 139.11
Beilstein Registry Number 118954
EC Number 212-859-0
MDL number MFCD00006294
PubChem Substance ID 24879992
Popular Documents: Specification Sheet (PDF) | FTIR (PDF)
| Related Categories | Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, E-H, Puriss p.a., Mass Spectrometry (MS), |
| grade | matrix substance for MALDI-MS |
| assay | ≥99.0% (HPLC) |
| mp | 213-218 °C(lit.) |
| cation traces | Ba: ≤5 mg/kg |
| Ca: ≤50 mg/kg | |
| Cd: ≤5 mg/kg | |
| Co: ≤5 mg/kg |
Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. 56197.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?