Synonym:
1,3-
CAS Number 5259-01-8
Empirical Formula (Hill Notation) C19H32O2
Molecular Weight 292.46
Beilstein Registry Number 2332964
MDL number MFCD16660790
| Related Categories | Alkylresorcinols, Analytical Standards, Analytical/Chromatography, Antioxidants, Chemical Class, |
| grade | analytical standard |
| assay | ≥95.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?