Synonym: Dideoxynucleoside Triphosphate Set
Popular Documents: Datasheet (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Mass Spectrometry (MS), Reagents for Mass Spectrometry (MS), Spectroscopy More... |
| grade | for MALDI MS |
| composition | ddATP, 10 mmol/L |
| ddCTP, 10 mmol/L | |
| ddGTP, 10 mmol/L | |
| ddTTP, 10 mmol/L | |
| storage temp. | −20°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?