EMAIL THIS PAGE TO A FRIEND

57594 Fluka

(+)-Menthofuran

technical, ≥90% (GC)

Synonym: (R)-3,6-Dimethyl-4,5,6,7-tetrahydrobenzofuran

  • CAS Number 17957-94-7

  • Empirical Formula (Hill Notation) C10H14O

  • Molecular Weight 150.22

  •  Beilstein Registry Number 2729

  •  MDL number MFCD00142332

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Furans, Heterocyclic Building Blocks More...
grade   technical
assay   ≥90% (GC)
optical activity   [α]/D +97±5°, neat
bp   204-206 °C(lit.)
density   0.97 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Price and Availability

Customers Also Viewed

(+)-Menthofuran

analytical standard

(<I>R</I>)-(+)-Pulegone

≥85%

<SC>L</SC>-Menthyl acetate

natural, ≥98%, FCC

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Pennyroyal toxicity: measurement of toxic metabolite levels in two cases and review of the literature. Anderson IB, Mullen WH, Meeker JE, et al. Ann. Intern. Med. 124(8), 726-34, (1996)

Loading...


Multiple organ failure after ingestion of pennyroyal oil from herbal tea in two infants. Bakerink JA, Gospe SM, Dimand RJ, et al. Pediatrics 98(5), 944-7, (1996)

Loading...


Effect of Mentha x piperita essential oil and monoterpenes on cucumber root membrane potential. Maffei M, Camusso W, and Sacco S Phytochemistry 58(5), 703-7, (2001)

Loading...


Menthofuran regulates essential oil biosynthesis in peppermint by controlling a downstream monoterpene reductase. Mahmoud SS and Croteau RB Proc. Natl. Acad. Sci. U. S. A. 100(24), 14481-6, (2003)

Loading...


A systems biology approach identifies the biochemical mechanisms regulating monoterpenoid essential oil composition in peppermint. Rios-Estepa R, Turner GW, Lee JM, et al. Proc. Natl. Acad. Sci. U. S. A. 105(8), 2818-23, (2008)

Loading...


Demonstration that menthofuran synthase of mint (Mentha) is a cytochrome P450 monooxygenase: cloning, functional expression, and characterization of the responsible gene. Bertea CM, Schalk M, Karp F, et al. Arch. Biochem. Biophys. 390(2), 279-86, (2001)

Loading...


In vitro inhibition of cytochrome P450 enzymes in human liver microsomes by a potent CYP2A6 inhibitor, trans-2-phenylcyclopropylamine (tranylcypromine), and its nonamine analog, cyclopropylbenzene. Taavitsainen P, Juvonen R, and Pelkonen O Drug Metab. Dispos. 29(3), 217-22, (2001)

Loading...


Metabolism of (R)-(+)-pulegone and (R)-(+)-menthofuran by human liver cytochrome P-450s: evidence for formation of a furan epoxide. Khojasteh-Bakht SC, Chen W, Koenigs LL, et al. Drug Metab. Dispos. 27(5), 574-80, (1999)

Loading...


(R)-(+)-Menthofuran is a potent, mechanism-based inactivator of human liver cytochrome P450 2A6. Khojasteh-Bakht SC, Koenigs LL, Peter RM, et al. Drug Metab. Dispos. 26(7), 701-4, (1998)

Loading...


In vivo studies on the metabolism of the monoterpene pulegone in humans using the metabolism of ingestion-correlated amounts (MICA) approach: explanation for the toxicity differences between (S)-(-)- and (R)-(+)-pulegone. Engel W J. Agric. Food Chem. 51(22), 6589-97, (2003)

Loading...


Biosynthesis of menthofuran in Mentha x piperita: stereoselective and mechanistic studies. Fuchs S, Zinn S, Beck T, et al. J. Agric. Food Chem. 47(10), 4100-5, (1999)

Loading...


Nitration versus nitrosation chemistry of menthofuran: remarkable fragmentation and dimerization pathways and expeditious entry into dehydromenthofurolactone. De Lucia M, Mainieri F, Verotta L, et al. J. Org. Chem. 72(26), 10123-9, (2007)

Loading...


Metabolism and toxicity of menthofuran in rat liver slices and in rats. Khojasteh SC, Oishi S, and Nelson SD Chem. Res. Toxicol. 23(11), 1824-32, (2010)

Loading...


Hepatoprotective effect of C-phycocyanin: protection for carbon tetrachloride and R-(+)-pulegone-mediated hepatotoxicty in rats. Vadiraja BB, Gaikwad NW, and Madyastha KM Biochem. Biophys. Res. Commun. 249(2), 428-31, (1998)

Loading...


Metabolism of (R)-(+)-menthofuran in Fischer-344 rats: identification of sulfonic acid metabolites. Chen LJ, Lebetkin EH, and Burka LT Drug Metab. Dispos. 31(10), 1208-13, (2003)

Loading...


Inhibition and inactivation of cytochrome P450 2A6 and cytochrome P450 2A13 by menthofuran, β-nicotyrine and menthol. Kramlinger VM, von Weymarn LB, and Murphy SE Chem. Biol. Interact. 197(2-3), 87-92, (2012)

Loading...


Characterization of rat liver proteins adducted by reactive metabolites of menthofuran. Khojasteh SC, Hartley DP, Ford KA, et al. Chem. Res. Toxicol. 25(11), 2301-9, (2012)

Loading...


Volatile organic compounds from rhizobacteria increase biosynthesis of essential oils and growth parameters in peppermint (Mentha piperita). Santoro MV, Zygadlo J, Giordano W, et al. Plant Physiol. Biochem. 49(10), 1177-82, (2011)

Loading...


Molecular response of Musca domestica L. to Mintostachys verticillata essential oil, (4R)+-pulegone and menthone. Rossi YE, Canavoso L, and Palacios SM Fitoterapia 83(2), 336-42, (2012)

Loading...


Chemical composition and toxicity of the essential oils from Cunila species (Lamiaceae) on the cattle tick Rhipicephalus (Boophilus) microplus. Apel MA, Ribeiro VL, Bordignon SA, et al. Parasitol. Res. 105(3), 863-8, (2009)

Loading...


Vapor toxicity of three prototype volatile insecticidal compounds to house fly (Diptera: Muscidae). Chaskopoulou A, Pereira RM, Scharf ME, et al. J. Med. Entomol. 46(6), 1400-6, (2009)

Loading...


Quenching of singlet molecular oxygen by Commiphora myrrha extracts and menthofuran. Racine P and Auffray B Fitoterapia 76(3-4), 316-23, (2005)

Loading...


Evaluation of inhibition selectivity for human cytochrome P450 2A enzymes. Stephens ES, Walsh AA, and Scott EE Drug Metab. Dispos. 40(9), 1797-802, (2012)

Loading...


14C-labeled pulegone and metabolites binding to alpha2u-globulin in kidneys of male F-344 rats. Ferguson LJ, Lebetkin EH, Lih FB, et al. J. Toxicol. Environ. Health A 70(17), 1416-23, (2007)

Loading...


In vitro production of M. × piperita not containing pulegone and menthofuran. Bertoli A, Leonardi M, Krzyzanowska J, et al. Acta Biochim. Pol. 59(3), 417-23, (2012)

Loading...


Beil. 17,V,1,518

Related Products

replaced by

Product #

Description

Add to Cart

63661 (+)-Menthofuran, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?