• USA Home
  • 57614 - Inulin from dahlia tubers

EMAIL THIS PAGE TO A FRIEND

57614 Fluka

Inulin from dahlia tubers

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrate adjuvants, Carbohydrates, Carbohydrates A to Z, Carbohydrates H-L,
optical activity   [α]20/D −37.0±2°, c = 6.5% in H2O
mol wt   Mr ~5000
ign. residue   ≤0.2%
loss   ~5% loss on drying

Description

Other Notes

Polysaccharide reserve of Compositae.; Inulin containing plants as alternative resources1; Suitable for the production of fructose syrups, review2; Potentials for non-food utilization of fructose and inulin, review3

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

from dahlia tuber

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. R.H.F. Beck, W. Praznik Starch/Staerke 38, 391, (1986)

2. M. Kiersten Process. Biochem. 15, 2, (1980)

3. A. Fuchs Starch/Staerke 39, 335, (1987)

An inactivated cell culture Japanese encephalitis vaccine (JE-ADVAX) formulated with delta inulin adjuvant provides robust heterologous protection against West Nile encephalitis via cross-protective memory B cells and neutralizing antibody. Petrovsky N, Larena M, Siddharthan V, et al. J. Virol. 87(18), 10324-33, (2013)

Loading...


Short-chain fatty acids stimulate angiopoietin-like 4 synthesis in human colon adenocarcinoma cells by activating peroxisome proliferator-activated receptor γ. Alex S, Lange K, Amolo T, et al. Mol. Cell. Biol. 33(7), 1303-16, (2013)

Loading...


Acute kidney injury: arterial spin labeling to monitor renal perfusion impairment in mice-comparison with histopathologic results and renal function. Hueper K, Gutberlet M, Rong S, et al. Radiology 270(1), 117-24, (2014)

Loading...


JE-ADVAX vaccine protection against Japanese encephalitis virus mediated by memory B cells in the absence of CD8(+) T cells and pre-exposure neutralizing antibody. Larena M, Prow NA, Hall RA, et al. J. Virol. 87(8), 4395-402, (2013)

Loading...


Tailor-made fructan synthesis in plants: a review. van Arkel J, Sévenier R, Hakkert JC, et al. Carbohydr. Polym. 93(1), 48-56, (2013)

Loading...


Impact of inulin and okara on Lactobacillus acidophilus La-5 and Bifidobacterium animalis Bb-12 viability in a fermented soy product and probiotic survival under in vitro simulated gastrointestinal conditions. Bedani R, Rossi EA, and Isay Saad SM Food Microbiol. 34(2), 382-9, (2013)

Loading...


Development of a dry, stable and inhalable acyl-homoserine-lactone-acylase powder formulation for the treatment of pulmonary Pseudomonas aeruginosa infections. Wahjudi M, Murugappan S, van Merkerk R, et al. Eur. J. Pharm. Sci. 48(4-5), 637-43, (2013)

Loading...


Unraveling protein stabilization mechanisms: vitrification and water replacement in a glass transition temperature controlled system. Grasmeijer N, Stankovic M, de Waard H, et al. Biochim. Biophys. Acta 1834(4), 763-9, (2013)

Loading...


Urinary creatinine-splitting bacteria after ileal-loop diversion causing underestimate of glomerular filtration rate. Seifter JL Am. J. Med. 127(12), e11-2, (2014)

Loading...


[Evaluating kidney function in family practice. Important - but difficult!]. Stiefelhagen P MMW Fortschr. Med. 155(3), 14-5, (2013)

Loading...


A morphological and functional comparison of proximal tubule cell lines established from human urine and kidney tissue. Jansen J, Schophuizen CM, Wilmer MJ, et al. Exp. Cell Res. 323(1), 87-99, (2014)

Loading...


Changes in intestinal morphology and microbiota caused by dietary administration of inulin and Bacillus subtilis in gilthead sea bream (Sparus aurata L.) specimens. Cerezuela R, Fumanal M, Tapia-Paniagua ST, et al. Fish Shellfish Immunol. 34(5), 1063-70, (2013)

Loading...


Influence of fat replacement by inulin on rheological properties, kinetics of rennet milk coagulation, and syneresis of milk gels. Arango O, Trujillo AJ, and Castillo M J. Dairy Sci. 96(4), 1984-96, (2013)

Loading...


Effects of dietary inulin, Bacillus subtilis and microalgae on intestinal gene expression in gilthead seabream (Sparus aurata L.). Cerezuela R, Meseguer J, and Esteban MÁ Fish Shellfish Immunol. 34(3), 843-8, (2013)

Loading...


Calcium and α-tocopherol suppress cured-meat promotion of chemically induced colon carcinogenesis in rats and reduce associated biomarkers in human volunteers. Pierre FH, Martin OC, Santarelli RL, et al. Am. J. Clin. Nutr. 98(5), 1255-62, (2013)

Loading...


Enzymatic synthesis of fructooligosaccharides by inulinases from Aspergillus niger and Kluyveromyces marxianus NRRL Y-7571 in aqueous-organic medium. Silva MF, Rigo D, Mossi V, et al. Food Chem. 138(1), 148-53, (2013)

Loading...


The interplay between fiber and the intestinal microbiome in the inflammatory response. Kuo SM Adv. Nutr. 4(1), 16-28, (2013)

Loading...


Creatinine- versus cystatine C-based equations in assessing the renal function of candidates for liver transplantation with cirrhosis. De Souza V, Hadj-Aissa A, Dolomanova O, et al. Hepatology 59(4), 1522-31, (2014)

Loading...


Fecal microbial communities of healthy adult dogs fed raw meat-based diets with or without inulin or yeast cell wall extracts as assessed by 454 pyrosequencing. Beloshapka AN, Dowd SE, Suchodolski JS, et al. FEMS Microbiol. Ecol. 84(3), 532-41, (2013)

Loading...


Yacon (Smallanthus sonchifolius): a functional food. Delgado GT, Tamashiro WM, Maróstica Junior MR, et al. Plant Foods Hum. Nutr. 68(3), 222-8, (2013)

Loading...


Application of poly(ethylene glycol)-b-poly(epsilon-caprolactone) copolymers with different Poly(ethylene glycol) contents for the preparation of PEG-coated nanoparticles. Hou J, Qian C, Zhang Y, et al. J. Biomed. Nanotechnol. 9(2), 231-7, (2013)

Loading...


Cloning of inulin fructotransferase (DFA III-producing) gene from Arthrobacter sp. L68-1. Haraguchi K Carbohydr. Polym. 93(2), 473-7, (2013)

Loading...


Changes in sensory properties and consumer acceptance of reduced fat pork Lyon-style and liver sausages containing inulin and citrus fiber as fat replacers. Tomaschunas M, Zörb R, Fischer J, et al. Meat Science 95(3), 629-40, (2013)

Loading...


Difructosan anhydrides III preparation from sucrose by coupled enzyme reaction. Hang H, Miao M, Li Y, et al. Carbohydr. Polym. 92(2), 1608-11, (2013)

Loading...


Preparation of inulin-type fructooligosaccharides using fast protein liquid chromatography coupled with refractive index detection. Li J, Cheong KL, Zhao J, et al. J. Chromatogr. A 1308, 52-7, (2013)

Loading...


Expression of an exoinulinase gene from Aspergillus ficuum in Escherichia coli and its characterization. Chen XM, Xu XM, Jin ZY, et al. Carbohydr. Polym. 92(2), 1984-90, (2013)

Loading...


Effects of konjac glucomannan, inulin and cellulose on acute colonic responses to genotoxic azoxymethane. Wu WT, Yang LC, and Chen HL Food Chem. 155, 304-10, (2014)

Loading...


Dry-fermented chicken sausage produced with inulin and corn oil: physicochemical, microbiological, and textural characteristics and acceptability during storage. Menegas LZ, Pimentel TC, Garcia S, et al. Meat Science 93(3), 501-6, (2013)

Loading...


Inulin-enriched pasta improves intestinal permeability and modifies the circulating levels of zonulin and glucagon-like peptide 2 in healthy young volunteers. Russo F, Linsalata M, Clemente C, et al. Nutr. Res. 32(12), 940-6, (2012)

Loading...


Ethanol production using whole plant biomass of Jerusalem artichoke by Kluyveromyces marxianus CBS1555. Kim S, Park JM, and Kim CH Appl. Biochem. Biotechnol. 169(5), 1531-45, (2013)

Loading...


Measurement of glomerular filtration rate during flight in a migratory bird using a single bolus injection of FITC-inulin. Gerson AR and Guglielmo CG Am. J. Physiol. Renal Physiol. 305(6), F823-9, (2013)

Loading...


Relationship between serum TSH levels and intrarenal hemodynamic parameters in euthyroid subjects. Tsuda A, Inaba M, Ichii M, et al. Eur. J. Endocrinol. 169(1), 45-50, (2013)

Loading...


Glutamine protects intestinal barrier function of colon epithelial cells from ethanol by modulating Hsp70 expression. Akagi R, Ohno M, Matsubara K, et al. Pharmacology 91(1-2), 104-11, (2013)

Loading...


Prediction of glomerular filtration rate in cancer patients by an equation for Japanese estimated glomerular filtration rate. Funakoshi Y, Fujiwara Y, Kiyota N, et al. Jpn. J. Clin. Oncol. 43(3), 271-7, (2013)

Loading...


The ABC transporter encoded at the pneumococcal fructooligosaccharide utilization locus determines the ability to utilize long- and short-chain fructooligosaccharides. Linke CM, Woodiga SA, Meyers DJ, et al. J. Bacteriol. 195(5), 1031-41, (2013)

Loading...


Effect of a synbiotic on infantile acute gastroenteritis. Gundogdu Z Benef. Microbes 4(3), 231-5, (2013)

Loading...


Identification and functional analysis of the gene cluster for fructan utilization in Prevotella intermedia. Fuse H, Fukamachi H, Inoue M, et al. Gene 515(2), 291-7, (2013)

Loading...


Synbiotic administration of canine-derived strain Lactobacillus fermentum CCM 7421 and inulin to healthy dogs. Strompfová V, Lauková A, and Cilik D Can. J. Microbiol. 59(5), 347-52, (2013)

Loading...


Evaluation of immune response elicited by inulin as an adjuvant with filarial antigens in mice model. Mahalakshmi N, Aparnaa R, and Kaliraj P Scand. J. Immunol. 80(4), 261-70, (2014)

Loading...


Characteristics of selected peripheral blood parameters in polar fox (Alopex lagopus L.) fed diets with inulin. Szymeczko R, Głowińska B, Burlikowska K, et al. Folia Biol. (Krakow.) 61(1-2), 113-8, (2013)

Loading...


Frequently asked questions of the Academy's knowledge center. Peregrin T J. Acad. Nutr. Diet. 113(7), 887-91, (2013)

Loading...


Tracking the microbiome functionality: focus on Propionibacterium species. Foligné B, Breton J, Mater D, et al. Gut 62(8), 1227-8, (2013)

Loading...


Prebiotics for obesity: a small light on the horizon? Tilg H and Gasbarrini A Gut 62(8), 1096-7, (2013)

Loading...


Merck 14,5004

Related Products

replaced by

Product #

Description

Add to Cart

I3754 Inulin from dahlia tubers

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?