EMAIL THIS PAGE TO A FRIEND

57971 Fluka

1-Iodooctane

purum, ≥97.0% (GC)

Synonym: Octyl iodide

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
grade   purum
assay   ≥97.0% (GC)
contains   silver wool as stabilizer
refractive index   n20/D 1.4878(lit.)
  n20/D 1.488
bp   225-226 °C(lit.)
mp   −46-−45 °C(lit.)
density   1.33 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

1-Bromooctane

99%

1-Iodobutane

99%

1-Iododecane

98%

1-Iodohexane

contains copper as stabilizer, ≥98%

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36
WGK Germany 
3
RTECS 
RG9700100
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 1,IV,425

Aldrich MSDS 1, 1101:B / Corp MSDS 1 (1), 2004:C / FT-IR 1 (1), 60:B / FT-IR 2 (1), 88:D / FT-NMR 1 (1), 88:C / NMR-Reference 2 (1), 63:D / RegBook 1 (1), 61:D / Structure Index 1, 7:A:7 / Vapor Phase 3, 88:A

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?