CAS Number 486-73-7
Empirical Formula (Hill Notation) C10H7NO2
Molecular Weight 173.17
Beilstein Registry Number 129177
EC Number 207-639-6
MDL number MFCD00006901
PubChem Substance ID 24881179
Popular Documents: FTNMR (PDF)
| Related Categories | Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Isoquinolines |
| grade | technical |
| assay | ≥97.0% (T) |
| mp | 164 °C (dec.)(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?