EMAIL THIS PAGE TO A FRIEND

59305 Fluka

2-Propanol

ACS reagent, for UV-spectroscopy, ≥99.8% (GC)

Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Purchase

Properties

grade   ACS reagent
  for UV-spectroscopy
vapor density   2.1 (vs air)
vapor pressure   33 mmHg ( 20 °C)
  44 mmHg ( 25 °C)
assay   ≥99.8% (GC)
autoignition temp.   750 °F
expl. lim.   2 %
refractive index   n20/D 1.377(lit.)
bp   82 °C(lit.)
mp   −89.5 °C(lit.)
density   0.785 g/mL at 25 °C(lit.)
λ   1 cm path, H2O reference
UV absorption   λ: 220 nm Amax: 0.4
  λ: 230 nm Amax: 0.12
  λ: 240 nm Amax: 0.05
  λ: 250 nm Amax: 0.02
  λ: 280 nm Amax: 0.004

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.





Customers Also Viewed

2-Propanol

LC-MS CHROMASOLV®

2-Propanol

CHROMASOLV®, for HPLC, 99.9%

2-Propanol

puriss. p.a., ACS reagent, ≥99.8% (GC)

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
7-16-24/25-26
RIDADR 
UN 1219 3 / PGII
WGK Germany 
1
RTECS 
NT8050000
Flash Point(F) 
53.6 °F
Flash Point(C) 
12 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Merck 14,5208

Beil. 1,IV,1461

Aldrich MSDS 1, 1542:C / Arctander, 2661 / Corp MSDS 1 (2), 2952:B / FT-IR 2 (1), 170:B / FT-IR 1 (1), 117:D / FT-NMR 1 (1), 175:B / Fenaroli / IR-Spectra (3), 70:B / IR-Spectra (2), 67:H / NMR-Reference 2 (1), 109:C / RegBook 61 (1), 129:A / RegBook 1 (1), 129:A / Sax 6, 2304 / Sax 6, 1652 / Sigma FT-IR 1 (1), 1342:B / Structure Index 1, 15:B:2 / Vapor Phase 3, 166:A

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?