Synonym:
2-
CAS Number 131-57-7
Linear Formula HOC6H3(OCH3)COC6H5
Molecular Weight 228.24
Beilstein Registry Number 1913145
EC Number 205-031-5
MDL number MFCD00008387
| Related Categories | Aldehydes and Ketones, Alphabetical, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | analytical standard |
| assay | ≥98% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| bp | 150-160 °C/5 mmHg(lit.) |
| mp | 62-64 °C(lit.) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?