Synonym: Polyoxyethylenesorbitan monooleate, Polysorbatum 80, TWEEN® 80
CAS Number 9005-65-6
MDL number MFCD00082107
Popular Documents: RAMAN FTIR (PDF)
| Related Categories | Cell Culture, Core Bioreagents, Detergents, Detergents N to Z, Detergents by Applications, |
| grade | Ph Eur |
| vapor pressure | <1 mmHg ( 20 °C) |
| description | non-ionic |
| pharmacopeia | testing & handling conforms to Pharmacopeia |
| mol wt | average mol wt 79,000 |
| aggregation number | 60 |
| refractive index |
n |
TWEEN is a registered trademark of Croda International PLC
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?