Synonym: Potassium nitrate solution
CAS Number 7757-79-1
MDL number MFCD00011409
PubChem Substance ID 24881778
| Related Categories | AAS, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, P, |
| shelf life | (limited shelf life, expiry date on the label) |
| concentration | in nitric acid |
| 1.000 g/L K+ |
prepared with KNO3 and HNO3 (~0.5 M)
Customers Also Viewed
TraceCERT®, 1000 mg/L K in nitric acid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?