60155 Fluka

Potassium citrate tribasic monohydrate

Ph Eur

DOWNLOAD MSDS (PDF)

Synonym: Citric acid tripotassium salt, Kalii citras, Tripotassium citrate

  • CAS Number 6100-05-6

  • Linear Formula HOC(COOK)(CH2COOK)2 · H2O

  • Molecular Weight 324.41

  •  Beilstein Registry Number 3924344

  •  EC Number 231-905-0

  •  MDL number MFCD00150442

  • Popular Documents:  RAMAN FTIR (PDF)  

Properties

Related Categories Biological Buffers, Pharmacopoeia, Pharmacopoeia A-Z, USP/MultiCompendial Grade Buffers
grade   Ph Eur
pharmacopeia   testing & handling conforms to Pharmacopeia
mp   275 °C (dec.)(lit.)

Price and Availability

Customers Also Viewed

Potassium citrate tribasic monohydrate

meets USP testing specifications

Potassium citrate tribasic monohydrate

eluent additive for LC-MS

Potassium citrate tribasic monohydrate

purum p.a., ≥99.0% (NT)

Potassium citrate tribasic monohydrate

meets analytical specification of Ph. Eur., BP, FCC, E332, 99-100.5% (calc with ref. to anhyd. subst.)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?