Synonym: Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate
CAS Number 7778-77-0
Linear Formula KH2PO4
Molecular Weight 136.09
EC Number 231-913-4
MDL number MFCD00011401
PubChem Substance ID 24881814
Popular Documents: RAMAN FTIR (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents, HPLC, |
| grade | for HPLC |
| description | anhydrous |
| assay | ≥99.5% (T) |
| ≥99.5% | |
| pKa (25 °C) | (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid) |
| mp | 252.6 °C(lit.) |
| solubility | H2O: soluble0.1 g/mL at 25 °C, clear, colorless |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?