CAS Number 17084-13-8
Linear Formula KPF6
Molecular Weight 184.06
EC Number 241-143-0
MDL number MFCD00011412
PubChem Substance ID 24881833
| Related Categories | Chemical Synthesis, Inorganic Salts, Potassium, Synthetic Reagents |
| grade | purum |
| assay | ≥98.0% (gravimetric) |
| mp | 575 °C(lit.) |
| solubility | H2O: slightly soluble |
| density | 2.75 g/mL at 25 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?