Synonym: cis-Tamoxifen-15N,N,N-dimethyl-13C2 solution
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, Drugs & Metabolites, |
| grade | drug standard |
| isotopic purity | 99 atom % 13C |
| 98 atom % 15N | |
| concentration | 1 mg/mL in methanol |
| mass shift | M+3 |
| storage temp. | −20°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?