60365 Fluka

Potassium L-tartrate monobasic

purum p.a., ≥99.0% (T)

DOWNLOAD MSDS (PDF)

Synonym: L-(+)-Tartaric acid monopotassium salt, Potassium hydrogen L-tartrate

  • CAS Number 868-14-4

  • Empirical Formula (Hill Notation) C4H5KO6

  • Molecular Weight 188.18

  •  Beilstein Registry Number 6119985

  •  EC Number 212-769-1

  •  MDL number MFCD00065392

  • Popular Documents:  FTIR (PDF)  

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acid Salts, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
grade   purum p.a.
assay   ≥99.0% (T)
optical activity   [α]20/D +32.0±0.5°, c = 10% in 1 M NaOH
pH   3.4-3.7 (25 °C, 0.5% in H2O)
anion traces   chloride (Cl-): ≤200 mg/kg
  sulfate (SO42-): ≤1000 mg/kg
cation traces   Ca: ≤100 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  Na: ≤1000 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability

Customers Also Viewed

Potassium <SC>L</SC>-tartrate monobasic

puriss., meets analytical specification of Ph.Eur., BP, FCC, 99.5-100.5% (related to dried substance)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?