60592 Fluka

Potassium tetraoxalate dihydrate

puriss. p.a., ≥99.5%

DOWNLOAD MSDS (PDF)

Synonym: Oxalic acid hemipotassium salt

  • CAS Number 6100-20-5

  • Linear Formula KH3(C2O4)2 · 2H2O

  • Molecular Weight 254.19

  •  Beilstein Registry Number 3854419

  •  EC Number 204-874-6

  •  MDL number MFCD00150443

  •  PubChem Substance ID 24881924

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Biochemicals and Reagents, Chelators,
grade   puriss. p.a.
assay   ≥99.5% (RT)
  ≥99.5%
anion traces   chloride (Cl-): ≤10 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  NH4+: ≤50 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?