Empirical Formula (Hill Notation) 13C2C24H2915NO
Molecular Weight 374.49
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, T, TA - TE More... |
| isotopic purity | 99 atom % 13C |
| 98 atom % 15N | |
| mass shift | M+3 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?