Synonym:
4-
CAS Number 100-10-7
Empirical Formula (Hill Notation) C9H11NO
Molecular Weight 149.19
Beilstein Registry Number 4132845
MDL number MFCD00003381
PubChem Substance ID 24882019
Popular Documents: Datasheet (PDF) | FTNMR (PDF)
| Related Categories | Aldehydes, Biochemical Reagents for Identification, Building Blocks, C9, Carbonyl Compounds, |
| grade | for microbiology |
| shelf life | (limited shelf life, expiry date on the label) |
| composition | n-butanol |
|
4-dimethylaminobenzaldehy | |
| hydrochloric acid | |
| refractive index |
n |
| storage temp. | 2-8°C |
In the presence of oxygen, some bacteria, like E. coli, are able to split tryptophan into indole and α-aminopropionic acid. This reagent is for detecting the indole and identifying the indole-positive and indole-negative microorganisms.
Solution of 4-(dimethylamino)benzalde
Customers Also Viewed
for microbiology
for microbiology
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?