CAS Number 28981-97-7
Empirical Formula (Hill Notation) C17H13ClN4
Molecular Weight 308.76
EC Number 200-659-6
MDL number MFCD00078881
PubChem Substance ID 24882050
| Related Categories | A, AA to AL, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard, for drug analysis |
| drug control | kontrollierte Droge in Deutschlandregulated under CDSA - not available from Sigma-Aldrich Canada |
| availability | not available in Japan |
| concentration | 1 mg/mL in methanol |
| storage temp. | −20°C |
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?