613339 Fluka

Formamide solution

NMR reference standard, 90% in DMSO-d6 (99.9 atom % D)

DOWNLOAD MSDS (PDF)
  • Empirical Formula (Hill Notation) CH3NO

  • Molecular Weight 45.04

  •  MDL number MFCD00007941

  •  PubChem Substance ID 24882117

Properties

Related Categories Alphabetical Index of Analytical Standards, Alphabetical Listings, Analytical Standards, Analytical/Chromatography, E-F,
grade   NMR reference standard
concentration   90% in DMSO-d6 (99.9 atom % D)
NMR tube O.D. × L   3 mm × 8 in.

Description

Features and Benefits

15N sensitivity

Price and Availability

Request a Quote for this
Stable Isotope Product


Isotec Stable IsotopesRequest Stable Isotope product pricing
and availablity.






Customers Also Viewed

Dimethyl sulfoxide-d<SUB>6</SUB>

"100%", 99.96 atom % D

Formamide

≥99.5% (GC), BioReagent, for molecular biology

Formamide

for molecular biology, ≥99.5% (GC), liquid

Formamide
295876Formamide

spectrophotometric grade, ≥99%

Formamide solution

NMR reference standard, 90% in DMSO-d6 (99.9 atom % D)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?