CAS Number 53857-91-3
MDL number MFCD01867336
| Related Categories | Analytical/Chromatography, Cation, Cation Exchange Media, LPLC Separation Media, LPLC/TLC/Paper Chromatography, |
| limit | 120 °C max. temp. (250 °F) |
| moisture | 53-55% |
| loss | 45-55% loss on drying, 110 °C |
| matrix | styrene-divinylbenzene (macroporous) |
| matrix active group | sulfonic acid functional group |
| uniform particle size | 660 μm std dev ±70 μm |
| operating pH | 0 - 14 |
Strongly acidic cation exchange resin especially suited for purification of condensates, demineralization of water at higher temperature, treatment of galvanic rinsing water, removal of metal ions from process solutions and passivating dips, and for the binding of heavy metals in hydrometallurgy.
Uniform particle size (monodispersed) provides optimized kinetics and leads to an increased capacity when compared to heterodispersed bead size distribution.
spherical particles with a special particle size distribution
Lewatit is a registered trademark of Lanxess Deutschland GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?