CAS Number 57285-14-0
MDL number MFCD01867337
| Related Categories | Analytical/Chromatography, Cation, Chelating Ion Exchange Media, LPLC Separation Media, LPLC/TLC/Paper Chromatography, |
| description | weakly acidic cation exchanger |
| form | particles (spherical) |
| moisture | 53-58% |
| loss | 45-60% loss on drying, 110 °C |
| matrix | styrene-divinylbenzene (macroporous) |
| matrix active group | iminodiacetic acid functional group |
| particle size | 400-1250 μm |
Chelating resin especially suited for the selective binding of heavy metal cations from weakly acidic to weakly basic solutions.
Lewatit is a registered trademark of Lanxess Deutschland GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?