• USA Home
  • 62150 - Lindlar Catalyst Poison

EMAIL THIS PAGE TO A FRIEND

62150 Fluka

Lindlar Catalyst Poison

Synonym: 2,2′-(Ethylenedithio)diethanol, 3,6-Dithiaoctane-1,8-diol, Lindlar Catalyst Poison

Purchase

Description

Other Notes

Efficient additive for selective hydrogenations over Lindlar-catalyst1

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. H. Lindlar, R. Dubuis Organic Synth. V, 880, (1975)

Beil. 1,IV,2450

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?