Synonym:
Linolelaidic acid methyl ester,
Methyl trans,trans-
CAS Number 2566-97-4
Empirical Formula (Hill Notation) C19H34O2
Molecular Weight 294.47
Beilstein Registry Number 1727615
EC Number 219-901-7
MDL number MFCD00069997
PubChem Substance ID 24882197
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Biochemicals and Reagents, Chromatography, |
| grade | analytical standard |
| assay | ≥99.0% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | −20°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?