62472 Fluka

Lithium carbonate

purum p.a., ≥98.0% (T)

DOWNLOAD MSDS (PDF)

Synonym: Carbolithium, Carbonic acid lithium salt

  • CAS Number 554-13-2

  • Linear Formula Li2CO3

  • Molecular Weight 73.89

  •  Beilstein Registry Number 3999191

  •  EC Number 209-062-5

  •  MDL number MFCD00011084

Properties

Related Categories Chemical Synthesis, Inorganic Bases, Synthetic Reagents More...
grade   purum p.a.
assay   ≥98.0% (T)
mp   618 °C(lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤1000 mg/kg
cation traces   Ca: ≤100 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤500 mg/kg
  Na: ≤1000 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability

Customers Also Viewed

Lithium carbonate

TraceSELECT®, for trace analysis, ≥99.998% (metals basis)

Lithium carbonate

BioXtra, ~99%

Lithium carbonate

puriss. p.a., ACS reagent, reagent (for microscopy), ≥99.0% (T)

Lithium carbonate

purum, ≥99%

Lithium carbonate

99.997% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?