62489 Fluka

Lithium dimethylamide

purum, ≥95.0%

DOWNLOAD MSDS (PDF)
  • CAS Number 3585-33-9

  • Linear Formula (CH3)2NLi

  • Molecular Weight 51.02

  •  Beilstein Registry Number 3618233

  •  EC Number 222-714-3

  •  MDL number MFCD00008295

  •  PubChem Substance ID 24882229

Properties

grade   purum
assay   ≥95.0%

Price and Availability

Customers Also Viewed

Dimethylamine

anhydrous, ≥99%

Lithium dimethylamide preparation

5 wt. % in hexanes

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?